missing translation for 'onlineSavingsMsg'
Learn More

1,1-Cyclobutanedicarboxylic acid, 99%

Catalog No. AAA1428822
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1428822 100 g
AAA1428806 5 g
AAA1428814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1428822 Supplier Thermo Scientific Chemicals Supplier No. A1428822
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 5445-51-2 | C6H8O4 | 144.126 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5445-51-2
Molecular Formula C6H8O4
Molecular Weight (g/mol) 144.126
MDL Number MFCD00001325
InChI Key CCQPAEQGAVNNIA-UHFFFAOYSA-N
Synonym 1,1-cyclobutanedicarboxylic acid, h2cbdca, 1,1-cyclobutanedicarboxylicacid, cyclobutanedicarboxylic acid, 1,1-cyclobutane dicarboxylic acid, 1,1-cyclobutanedicarboxylate, cyclobutanedicarbonic acid, zlchem 254, pubchem14151, acmc-1am73
PubChem CID 2568
ChEBI CHEBI:35691
IUPAC Name cyclobutane-1,1-dicarboxylic acid
SMILES C1CC(C1)(C(=O)O)C(=O)O

Specifications

Melting Point ∼158°C (decomposition)
Quantity 100 g
UN Number UN3261
Beilstein 2046031
Formula Weight 144.13
Percent Purity 99%
Chemical Name or Material 1,1-Cyclobutanedicarboxylic acid
Hazard Category H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
EINECSNumber 226-651-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only