missing translation for 'onlineSavingsMsg'
Learn More

1,1-Cyclobutanedicarboxylic acid, 99%, Thermo Scientific™

Catalog No. AC110920250
Change view
Click to view available options
25g; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AC110920250 25g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC110920250 Supplier Thermo Scientific Chemicals Supplier No. 110920250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

CAS 5445-51-2
CAS Min % 98.5
CAS Max % 100.0
Loss on Drying 0.1% max. (50°C)
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Molecular Formula C6H8O4
Linear Formula C4H6(CO2H)2
Quantity 25g
MDL Number MFCD00001325
Beilstein 09, 724
Synonym 1,1-cyclobutanedicarboxylic acid, h2cbdca, 1,1-cyclobutanedicarboxylicacid, cyclobutanedicarboxylic acid, 1,1-cyclobutane dicarboxylic acid, 1,1-cyclobutanedicarboxylate, cyclobutanedicarbonic acid, zlchem 254, pubchem14151, acmc-1am73
InChI Key CCQPAEQGAVNNIA-UHFFFAOYSA-N
SMILES C1CC(C1)(C(=O)O)C(=O)O
IUPAC Name cyclobutane-1,1-dicarboxylic acid
Molecular Weight (g/mol) 144.13
PubChem CID 2568
ChEBI CHEBI:35691
Formula Weight 144.13
Clarity (5 w/v% aq. soln.) clear
Percent Purity 99%
Chemical Name or Material 1, 1-Cyclobutanedicarboxylic acid, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
EINECSNumber 226-651-2
TSCA TSCA

RUO – Research Use Only