missing translation for 'onlineSavingsMsg'
Learn More

1,1'-Carbonyldiimidazole 97.0+%, TCI America™

Catalog No. c01195g
Change view
Click to view available options
Quantity:
5 g
25 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C01195G 5 g
C011925G 25 g
C0119250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. C01195G Supplier TCI America Supplier No. C01195G
Only null left

Chemical Identifiers

CAS 530-62-1
Molecular Formula C7H6N4O
Molecular Weight (g/mol) 162.15
MDL Number MFCD00005286
InChI Key PFKFTWBEEFSNDU-UHFFFAOYSA-N
Synonym 1,1'-carbonyldiimidazole, n,n'-carbonyldiimidazole, carbonyldiimidazole, di 1h-imidazol-1-yl methanone, n,n-carbonyldiimidazole, 1h-imidazole, 1,1'-carbonylbis, 1,1'-carbonylbis-1h-imidazole, carbonyl diimidazole, diimidazol-1-yl ketone, 1,1-carbonyldiimidazole
PubChem CID 68263
IUPAC Name 1-(1H-imidazole-1-carbonyl)-1H-imidazole
SMILES O=C(N1C=CN=C1)N1C=CN=C1

Specifications

Melting Point 118°C
Color White-Yellow
pH 8
Quantity 5 g
UN Number 1759
Formula Weight 162.15
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1,1′-Carbonyldiimidazole [Coupling Agent for Peptides Synthesis]
EINECSNumber (5)-6491
TSCA Yes
Recommended Storage Refrigerator