missing translation for 'onlineSavingsMsg'
Learn More

1,1-Bis(morpholino)ethylene 97.0+%, TCI America™

Catalog No. b118010g
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B118010G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B118010G Supplier TCI America Supplier No. B118010G

Chemical Identifiers

CAS 14212-87-4
Molecular Formula C10H18N2O2
Molecular Weight (g/mol) 198.266
MDL Number MFCD00059775
InChI Key MNEZCVVWTRIDOE-UHFFFAOYSA-N
Synonym 1,1-Dimorpholinoethylene, 4,4′C-(Ethylene-1,1-diyl)bismorpholine
PubChem CID 543713
IUPAC Name 4-(1-morpholin-4-ylethenyl)morpholine
SMILES C=C(N1CCOCC1)N2CCOCC2

Specifications

Color White-Yellow
Quantity 10 g
Formula Weight 198.27
Percent Purity ≥97.0% (GC,T)
Physical Form Crystals
Chemical Name or Material 1,1-Bis(morpholino)ethylene