missing translation for 'onlineSavingsMsg'
Learn More

1,1-Bis(methylthio)ethylene 90.0+%, TCI America™

Catalog No. p-7119184
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B16501G 1 g
B16505G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B16501G Supplier TCI America Supplier No. B16501G

Chemical Identifiers

CAS 51102-74-0
Molecular Formula C4H8S2
Molecular Weight (g/mol) 120.23
MDL Number MFCD00191617
InChI Key GGCGAESAURTGKY-UHFFFAOYSA-N
Synonym Ketene Dimethyl Thioacetal
PubChem CID 142813
IUPAC Name 1,1-bis(methylsulfanyl)ethene
SMILES CSC(=C)SC

Specifications

Boiling Point 110°C
Quantity 1 g
Formula Weight 120.23
Percent Purity ≥90.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,1-Bis(methylthio)ethylene
TSCA No
Recommended Storage Refrigerator