missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,1-Bis(hydroxymethyl)cyclopropane, tech. 90%, Thermo Scientific™
$63.58 - $205.75
Chemical Identifiers
| CAS | 39590-81-3 |
|---|---|
| Molecular Formula | C5H10O2 |
| Molecular Weight (g/mol) | 102.133 |
| MDL Number | MFCD00464207 |
| InChI Key | YAINYZJQSQEGND-UHFFFAOYSA-N |
| Synonym | 1,1-bis hydroxymethyl cyclopropane, 1-hydroxymethyl cyclopropyl methanol, 1-hydroxymethylcyclopropyl methanol, 1,1-cyclopropanedimethanol, cyclopropane-1,1-diyldimethanol, 1,1-dimetholcyclopropane, 1,1-cyclopropyldimethanol, cyclopropanedimethanol, pubchem15258, acmc-209j6f |
| PubChem CID | 4101437 |
| IUPAC Name | [1-(hydroxymethyl)cyclopropyl]methanol |
| SMILES | C1CC1(CO)CO |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH5608303
|
Thermo Scientific Chemicals
H5608303 |
1 g |
Each for $63.58
|
|
|||||
|
AAH5608306
|
Thermo Scientific Chemicals
H5608306 |
5 g |
Each for $205.75
|
|
|||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 39590-81-3 | |
| 102.133 | |
| YAINYZJQSQEGND-UHFFFAOYSA-N | |
| 4101437 | |
| C1CC1(CO)CO |
| C5H10O2 | |
| MFCD00464207 | |
| 1,1-bis hydroxymethyl cyclopropane, 1-hydroxymethyl cyclopropyl methanol, 1-hydroxymethylcyclopropyl methanol, 1,1-cyclopropanedimethanol, cyclopropane-1,1-diyldimethanol, 1,1-dimetholcyclopropane, 1,1-cyclopropyldimethanol, cyclopropanedimethanol, pubchem15258, acmc-209j6f | |
| [1-(hydroxymethyl)cyclopropyl]methanol |
Specifications
| 39590-81-3 | |
| 235°C to 236°C | |
| C5H10O2 | |
| MFCD00464207 | |
| 1,1-bis hydroxymethyl cyclopropane, 1-hydroxymethyl cyclopropyl methanol, 1-hydroxymethylcyclopropyl methanol, 1,1-cyclopropanedimethanol, cyclopropane-1,1-diyldimethanol, 1,1-dimetholcyclopropane, 1,1-cyclopropyldimethanol, cyclopropanedimethanol, pubchem15258, acmc-209j6f | |
| C1CC1(CO)CO | |
| 102.133 | |
| 102.13 | |
| Technical |
| 1.065 | |
| 110°C (230°F) | |
| 1.47 | |
| 1 g | |
| YAINYZJQSQEGND-UHFFFAOYSA-N | |
| [1-(hydroxymethyl)cyclopropyl]methanol | |
| 4101437 | |
| 90% | |
| 1,1-Bis(hydroxymethyl)cyclopropane |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280i-P304+P340-P305+P351+P338-P312-P501c
H319
TSCA : No
Recommended Storage : Ambient temperatures