missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,1-Bis(hydroxymethyl)cyclopropane, 90%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 426250050
| Quantity | 5g |
|---|---|
| Packaging | Glass bottle |
Chemical Identifiers
| 39590-81-3 | |
| 102.13 | |
| YAINYZJQSQEGND-UHFFFAOYSA-N | |
| 4101437 | |
| C1CC1(CO)CO |
| C5H10O2 | |
| MFCD00464207 | |
| 1,1-bis hydroxymethyl cyclopropane, 1-hydroxymethyl cyclopropyl methanol, 1-hydroxymethylcyclopropyl methanol, 1,1-cyclopropanedimethanol, cyclopropane-1,1-diyldimethanol, 1,1-dimetholcyclopropane, 1,1-cyclopropyldimethanol, cyclopropanedimethanol, pubchem15258, acmc-209j6f | |
| [1-(hydroxymethyl)cyclopropyl]methanol |
Specifications
| 1, 1-Bis(hydroxymethyl)cyclopropane | |
| 88.0 | |
| 1.0650g/mL | |
| 110°C | |
| Glass bottle | |
| 5g | |
| 1.065 | |
| YAINYZJQSQEGND-UHFFFAOYSA-N | |
| [1-(hydroxymethyl)cyclopropyl]methanol | |
| 4101437 | |
| 90% |
| 39590-81-3 | |
| 100.0 | |
| 215.0°C to 233.0°C | |
| 90% | |
| C5H10O2 | |
| MFCD00464207 | |
| 1,1-bis hydroxymethyl cyclopropane, 1-hydroxymethyl cyclopropyl methanol, 1-hydroxymethylcyclopropyl methanol, 1,1-cyclopropanedimethanol, cyclopropane-1,1-diyldimethanol, 1,1-dimetholcyclopropane, 1,1-cyclopropyldimethanol, cyclopropanedimethanol, pubchem15258, acmc-209j6f | |
| C1CC1(CO)CO | |
| 102.13 | |
| 102.13 |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
GHS P Statement
Wear eye protection/face protection.
Wash face,hands and any exposed skin thoroughly after handling.
If eye irritation persists: Get medical advice/attention.
GHS Signal Word: Warning