missing translation for 'onlineSavingsMsg'
Learn More

[1,1'-Bis(diphenylphosphino)ferrocene]palladium(II) Dichloride Dichloromethane Adduct 98.0+%, TCI America™

Catalog No. p-7119455
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B20641G 1 g
B20645G 5 g
B206425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B20641G Supplier TCI America Supplier No. B20641G
Only null left

Chemical Identifiers

CAS 95464-05-4
Molecular Formula C35H30Cl4FeP2Pd
Molecular Weight (g/mol) 816.64
MDL Number MFCD00792899
IUPAC Name Bis(diphenylphosphino)ferrocene]palladium(II) chloride
SMILES [Fe].ClCCl.Cl[Pd]Cl.c1ccc(c1)P(C1=CC=CC=C1)C1=CC=CC=C1.c1ccc(c1)P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 268°C
Quantity 1 g
Synonym [1,1′C-Bis(diphenylphosphino)ferrocene]dichloropalladium(II), Dichloro[1,1′C-bis(diphenylphosphino)ferrocene]palladium(II)
Formula Weight 816.64
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material [1,1′-Bis(diphenylphosphino)ferrocene]palladium(II) Dichloride Dichloromethane Adduct