missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

[1,1′-Bis(diphenylphosphino)ferrocene]nickel(II) Dichloride, TCI America™

$82.50 - $247.50

Chemical Identifiers

CAS 67292-34-6
Molecular Formula C34H28Cl2FeNiP2
MDL Number MFCD00270284
Synonym [1,1′-Bis(diphenylphosphino)ferrocene]dichloronickel(II), Dichloro[1,1′-bis(diphenylphosphino)ferrocene]nickel(II)
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B22261G View Documents TCI America
B22261G
1g Each for $82.50
 
B22265G View Documents TCI America
B22265G
5g Each for $247.50
 
Specifications

Chemical Identifiers

67292-34-6
C34H28Cl2FeNiP2
MFCD00270284
[1,1′-Bis(diphenylphosphino)ferrocene]dichloronickel(II), Dichloro[1,1′-bis(diphenylphosphino)ferrocene]nickel(II)

Specifications

Undesignated
1g
C34H28Cl2FeNiP2
[1,1′C-Bis(diphenylphosphino)ferrocene]dichloronickel(II), Dichloro[1,1′C-bis(diphenylphosphino)ferrocene]nickel(II)
Crystal-Powder at 20°C
[1,1′-Bis(diphenylphosphino)ferrocene]nickel(II) Dichloride
259°C
67292-34-6
MFCD00270284
683.98
≥97.0% (T)
Safety and Handling

Safety and Handling

TSCA : No