missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sigma Aldrich [1,1'-Bis(Diphenylphosphino)Ferrocene]Dichloropalladium(II)
Description
[1,1'-Bis(Diphenylphosphino)Ferrocene]Dichloropalladium(II)
Specifications
Specifications
| CAS | 72287-26-4 |
| Melting Point | 266°C to 283°C |
| Molecular Formula | C34H28Cl2FeP2Pd |
| Linear Formula | (C17H14P)2Fe -+ PdCl2 |
| MDL Number | MFCD00015757 |
| Quantity | 5 g |
| Synonym | Pd(dppf)Cl2 |
| Molecular Weight (g/mol) | 731.7 g/mol |