missing translation for 'onlineSavingsMsg'
Learn More

Sigma Aldrich [1,1'-Bis(Diphenylphosphino)Ferrocene]Dichloropalladium(II), Complex With Dichloromethane

Catalog No. 37967025G
Encompass
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
37967025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 37967025G Supplier Sigma Aldrich Supplier No. 37967025G
Only null left

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

[1,1'-Bis(Diphenylphosphino)Ferrocene]Dichloropalladium(II), Complex With Dichloromethane

Specifications

CAS 95464-05-4
Melting Point 275°C to 280°C
Molecular Formula C35H30Cl4FeP2Pd
Linear Formula C35H30Cl4FeP2Pd
MDL Number MFCD00792899
Quantity 25 g
Synonym Pd(dppf)Cl2 -+ CH2Cl2; Pd(dppf)Cl2 -+ DCM; [1,1'-Bis(diphenylphosphino)ferrocene]dichloropalladium(II) (1:1)
Molecular Weight (g/mol) 816.64 g/mol