missing translation for 'onlineSavingsMsg'
Learn More

[1,1'-Bis(diphenylphosphino)ferrocene]cobalt(II) Dichloride 98.0+%, TCI America™

Catalog No. p-7122907
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B33741G 1 g
B33745G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B33741G Supplier TCI America Supplier No. B33741G
Only null left

Chemical Identifiers

CAS 67292-36-8
Molecular Formula C34H28Cl2CoFeP2
Molecular Weight (g/mol) 684.22
MDL Number MFCD03427161
IUPAC Name [1,1'-Bis(diphenylphosphino)ferrocene]cobalt(II) chloride
SMILES [Fe].Cl[Co]Cl.c1ccc(c1)P(C1=CC=CC=C1)C1=CC=CC=C1.c1ccc(c1)P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 224°C
Color Green-Yellow
Quantity 1 g
Synonym [1,1′C-Bis(diphenylphosphino)ferrocene]dichlorocobalt(II), Dichloro[1,1′C-bis(diphenylphosphino)ferrocene]cobalt(II)
Formula Weight 684.22
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material [1,1′-Bis(diphenylphosphino)ferrocene]cobalt(II) Dichloride