missing translation for 'onlineSavingsMsg'
Learn More

1,1'-Bis(diisopropylphosphino)ferrocene 98.0+%, TCI America™

Catalog No. B2710100MG
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B2710100MG 100 mg
B27101G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B2710100MG Supplier TCI America Supplier No. B2710100MG

Chemical Identifiers

CAS 97239-80-0
Molecular Formula C22H36FeP2
Molecular Weight (g/mol) 418.32
MDL Number MFCD02684559
Synonym Bis[(diisopropylphosphino)cyclopentadienyl]iron
IUPAC Name 1,1'-Bis(diisopropylphosphino)ferrocene
SMILES [Fe].CC(C)P(C(C)C)c1cccc1.CC(C)P(C(C)C)c1cccc1

Specifications

Melting Point 50°C
Quantity 100 mg
Formula Weight 418.32
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1,1′-Bis(diisopropylphosphino)ferrocene