missing translation for 'onlineSavingsMsg'
Learn More

1,1'-Bis(di-tert-butylphosphino)ferrocene 95.0+%, TCI America™

Catalog No. B2711100MG
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B2711100MG 100 mg
B27111G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B2711100MG Supplier TCI America Supplier No. B2711100MG

Chemical Identifiers

CAS 84680-95-5
Molecular Formula C26H44FeP2
Molecular Weight (g/mol) 474.43
MDL Number MFCD01630818
InChI Key DFRHTHSZMBROSH-UHFFFAOYSA-N
Synonym Bis[(di-tert-butylphosphino)cyclopentadienyl]iron, dtbpf
IUPAC Name 1,1'-Bis(di-tert-butylphosphino)ferrocene
SMILES [Fe].CC(C)(C)P(c1cccc1)C(C)(C)C.CC(C)(C)P(c1cccc1)C(C)(C)C

Specifications

Melting Point 182°C
Quantity 100 mg
Formula Weight 474.43
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1,1′-Bis(di-tert-butylphosphino)ferrocene