missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

[1,1′-Bis(di-tert-butylphosphino)ferrocene]palladium(II) Dichloride, TCI America™

$122.22 - $421.67

Chemical Identifiers

CAS 95408-45-0
Molecular Formula C26H44Cl2FeP2Pd
Synonym Dichloro[1,1′C-bis(di-tert-butylphosphino)ferrocene]palladium(II), [1,1′C-Bis(di-tert-butylphosphino)ferrocene]dichloropalladium(II)
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B31601G View Documents TCI America
B31601G
1g Each for $122.22
 
B31605G View Documents TCI America
B31605G
5g Each for $421.67
 
Specifications

Chemical Identifiers

95408-45-0
C26H44Cl2FeP2Pd
Dichloro[1,1′C-bis(di-tert-butylphosphino)ferrocene]palladium(II), [1,1′C-Bis(di-tert-butylphosphino)ferrocene]dichloropalladium(II)

Specifications

Undesignated
95408-45-0
Dichloro[1,1′C-bis(di-tert-butylphosphino)ferrocene]palladium(II), [1,1′C-Bis(di-tert-butylphosphino)ferrocene]dichloropalladium(II)
Crystal-Powder at 20°C
[1,1′-Bis(di-tert-butylphosphino)ferrocene]palladium(II) Dichloride
1g
C26H44Cl2FeP2Pd
651.75
≥98.0% (T)
Safety and Handling

Safety and Handling

TSCA : No