missing translation for 'onlineSavingsMsg'
Learn More

[1,1'-Bis(di-tert-butylphosphino)ferrocene]palladium(II) Dichloride 98.0+%, TCI America™

Catalog No. B31605G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B31605G 5 g
B31601G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B31605G Supplier TCI America Supplier No. B31605G
Only null left

Chemical Identifiers

CAS 95408-45-0
Molecular Formula C26H44Cl2FeP2Pd
Molecular Weight (g/mol) 651.75
MDL Number MFCD08064219
InChI Key DFRHTHSZMBROSH-UHFFFAOYSA-N
Synonym Dichloro[1,1′C-bis(di-tert-butylphosphino)ferrocene]palladium(II), [1,1′C-Bis(di-tert-butylphosphino)ferrocene]dichloropalladium(II)
IUPAC Name Dichloro[1,1'-bis(di-tert-butylphosphino)ferrocene]palladium(II)
SMILES [Fe].Cl[Pd]Cl.CC(C)(C)P(c1cccc1)C(C)(C)C.CC(C)(C)P(c1cccc1)C(C)(C)C

Specifications

Quantity 5 g
Formula Weight 651.75
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material [1,1′-Bis(di-tert-butylphosphino)ferrocene]palladium(II) Dichloride