missing translation for 'onlineSavingsMsg'
Learn More

1,1-Bis[4-[N,N-di(p-tolyl)amino]phenyl]cyclohexane 98.0+%, TCI America™

Catalog No. B20791G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B20791G 1 g
B20795G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B20791G Supplier TCI America Supplier No. B20791G
Only null left

Chemical Identifiers

CAS 58473-78-2
Molecular Formula C46H46N2
Molecular Weight (g/mol) 626.888
MDL Number MFCD03844768
InChI Key ZOKIJILZFXPFTO-UHFFFAOYSA-N
PubChem CID 94071
IUPAC Name 4-methyl-N-[4-[1-[4-(4-methyl-N-(4-methylphenyl)anilino)phenyl]cyclohexyl]phenyl]-N-(4-methylphenyl)aniline
SMILES CC1=CC=C(C=C1)N(C2=CC=C(C=C2)C)C3=CC=C(C=C3)C4(CCCCC4)C5=CC=C(C=C5)N(C6=CC=C(C=C6)C)C7=CC=C(C=C7)C

Specifications

Melting Point 186°C
Quantity 1 g
Synonym 4,4′C-Cyclohexylidenebis[N,N-di(p-tolyl)aniline], TAPC
Formula Weight 626.89
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material 1,1-Bis[4-[N,N-di(p-tolyl)amino]phenyl]cyclohexane