missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,1'-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)ferrocene 98.0+%, TCI America™
$268.43 - $1071.83
Chemical Identifiers
| CAS | 737776-93-1 |
|---|---|
| Molecular Formula | C22H32B2FeO4 |
| MDL Number | MFCD06411336 |
Chemical Identifiers
| 737776-93-1 | |
| C22H32B2FeO4 | |
| MFCD06411336 |
Specifications
| 737776-93-1 | |
| C22H32B2FeO4 | |
| 1 g | |
| 437.96 | |
| Crystalline Powder |
| 173°C | |
| MFCD06411336 | |
| 1,1′C-Ferrocenediboronic Acid Bis(pinacol) Ester | |
| ≥98.0% (GC,T) | |
| 1,1′-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)ferrocene |
Safety and Handling
TSCA : No