missing translation for 'onlineSavingsMsg'
Learn More

1,1'-[Biphenyl-4,4'-diylbis(methylene)]bis(4,4'-bipyridinium) Bis(hexafluorophosphate) 98.0+%, TCI America™

Catalog No. b2280500mg
Change view
Click to view available options
Quantity:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B2280500MG 500 mg
B2280100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B2280500MG Supplier TCI America Supplier No. B2280500MG

Chemical Identifiers

CAS 134815-78-4
Molecular Formula C34H28F12N4P2
Molecular Weight (g/mol) 782.55
MDL Number MFCD06797055
InChI Key ZRSWFWKWQFGSDD-UHFFFAOYSA-N
PubChem CID 11735404
IUPAC Name 1-{[4'-({[4,4'-bipyridin]-1-ium-1-yl}methyl)-[1,1'-biphenyl]-4-yl]methyl}-[4,4'-bipyridin]-1-ium; bis(hexafluoro-λ⁵-phosphanuide)
SMILES F[P-](F)(F)(F)(F)F.F[P-](F)(F)(F)(F)F.C(C1=CC=C(C=C1)C1=CC=C(C[N+]2=CC=C(C=C2)C2=CC=NC=C2)C=C1)[N+]1=CC=C(C=C1)C1=CC=NC=C1

Specifications

Melting Point 154°C
Quantity 500 mg
Formula Weight 782.55
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 1,1′-[Biphenyl-4,4′-diylbis(methylene)]bis(4,4′-bipyridinium) Bis(hexafluorophosph