missing translation for 'onlineSavingsMsg'
Learn More

1,1'-Binaphthyl 98.0+%, TCI America™

Catalog No. B08051G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B08051G 1 g
B08055G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B08051G Supplier TCI America Supplier No. B08051G

Chemical Identifiers

CAS 604-53-5
Molecular Formula C20H14
Molecular Weight (g/mol) 254.33
MDL Number MFCD00041740
InChI Key ZDZHCHYQNPQSGG-UHFFFAOYSA-N
Synonym 1,1'-binaphthalene, 1,1'-binaphthyl, 1,1'-dinaphthyl, binaphthyl, binaphthalene, 1-naphthalen-1-yl naphthalene, 1,1'-binaphtyl, di-.alpha.-naphthol, 1-1-naphthyl naphthalene, di-alpha-naphthol
PubChem CID 11789
IUPAC Name 1,1'-binaphthalene
SMILES C1=CC2=CC=CC(C3=CC=CC4=CC=CC=C34)=C2C=C1

Specifications

Color White-Yellow
Quantity 1 g
Formula Weight 254.33
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,1′-Binaphthyl