missing translation for 'onlineSavingsMsg'
Learn More

1,1'-Binaphthyl, 98%

Catalog No. p-7023372
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1685203 1 g
AAA1685206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1685203 Supplier Thermo Scientific Chemicals Supplier No. A1685203
Only null left

CAS: 604-53-5 | C20H14 | 254.33 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 604-53-5
Molecular Formula C20H14
Molecular Weight (g/mol) 254.33
MDL Number MFCD00041740
InChI Key ZDZHCHYQNPQSGG-UHFFFAOYSA-N
Synonym 1,1'-binaphthalene, 1,1'-binaphthyl, 1,1'-dinaphthyl, binaphthyl, binaphthalene, 1-naphthalen-1-yl naphthalene, 1,1'-binaphtyl, di-.alpha.-naphthol, 1-1-naphthyl naphthalene, di-alpha-naphthol
PubChem CID 11789
IUPAC Name 1-naphthalen-1-ylnaphthalene
SMILES C1=CC2=CC=CC(C3=CC=CC4=CC=CC=C34)=C2C=C1

Specifications

Melting Point 144°C to 146°C
Quantity 1 g
Formula Weight 254.33
Percent Purity 98%
Chemical Name or Material 1,1'-Binaphthyl
EINECSNumber 210-070-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only