missing translation for 'onlineSavingsMsg'
Learn More

(±)-1,1'-Bi(2-naphthylamine), 97%

CAS: 4488-22-6 | C20H16N2 | 284.362 g/mol

$181.82 - $580.78

Chemical Identifiers

CAS 4488-22-6
Molecular Formula C20H16N2
Molecular Weight (g/mol) 284.362
MDL Number MFCD00145204
InChI Key DDAPSNKEOHDLKB-UHFFFAOYSA-N
Synonym 1,1'-binaphthalene-2,2'-diamine, r-1,1'-binaphthalene-2,2'-diamine, s---1,1'-binaphthyl-2,2'-diamine, 1,1'-binaphthyl-2,2'-diamine, s---2,2'-diamino-1,1'-binaphthalene, 2,2'-diamino-1,1'-dinaphthyl, 1,1'-bi 2-naphthylamine, 1,1'-bi 2-naphthalenylamine, binam, r-+-2,2'-diamino-1,1'-binaphthalene
PubChem CID 20571
IUPAC Name 1-(2-aminonaphthalen-1-yl)naphthalen-2-amine
SMILES C1=CC=C2C(=C1)C=CC(=C2C3=C(C=CC4=CC=CC=C43)N)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH3126403
View Documents
Thermo Scientific Chemicals
H3126403
1 g N/A
Only null left
 
AAH3126406
View Documents
Thermo Scientific Chemicals
H3126406
5 g N/A
Only null left
 
Description

Description

Novel axially chiral Rh N-heterocyclic carbene complexes were prepared from axially dissymmetric 1,1?-binaphthalenyl-2,2?-diamine and applied in the Rh-catalyzed enantioselective hydrosilylation of methyl ketones. The enantioselective direct aldol reaction, organocatalyzed by recoverable BINAM- prolinamide derivatives can be highly accelerated by a catalytic amount of a carboxylic acid without a detrimental of the obtained enantioselectivities.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Novel axially chiral Rh N-heterocyclic carbene complexes were prepared from axially dissymmetric 1,1′-binaphthalenyl-2,2′-diamine and applied in the Rh-catalyzed enantioselective hydrosilylation of methyl ketones. The enantioselective direct aldol reaction, organocatalyzed by recoverable BINAM- prolinamide derivatives can be highly accelerated by a catalytic amount of a carboxylic acid without a detrimental of the obtained enantioselectivities.

Solubility
Insoluble in water

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications

Chemical Identifiers

4488-22-6
284.362
DDAPSNKEOHDLKB-UHFFFAOYSA-N
20571
C1=CC=C2C(=C1)C=CC(=C2C3=C(C=CC4=CC=CC=C43)N)N
C20H16N2
MFCD00145204
1,1'-binaphthalene-2,2'-diamine, r-1,1'-binaphthalene-2,2'-diamine, s---1,1'-binaphthyl-2,2'-diamine, 1,1'-binaphthyl-2,2'-diamine, s---2,2'-diamino-1,1'-binaphthalene, 2,2'-diamino-1,1'-dinaphthyl, 1,1'-bi 2-naphthylamine, 1,1'-bi 2-naphthalenylamine, binam, r-+-2,2'-diamino-1,1'-binaphthalene
1-(2-aminonaphthalen-1-yl)naphthalen-2-amine

Specifications

4488-22-6
C20H16N2
1 g
Insoluble in water
C1=CC=C2C(=C1)C=CC(=C2C3=C(C=CC4=CC=CC=C43)N)N
284.362
284.36
(±)-1,1'-Bi(2-naphthylamine)
188°C to 193°C
MFCD00145204
1,1'-binaphthalene-2,2'-diamine, r-1,1'-binaphthalene-2,2'-diamine, s---1,1'-binaphthyl-2,2'-diamine, 1,1'-binaphthyl-2,2'-diamine, s---2,2'-diamino-1,1'-binaphthalene, 2,2'-diamino-1,1'-dinaphthyl, 1,1'-bi 2-naphthylamine, 1,1'-bi 2-naphthalenylamine, binam, r-+-2,2'-diamino-1,1'-binaphthalene
DDAPSNKEOHDLKB-UHFFFAOYSA-N
1-(2-aminonaphthalen-1-yl)naphthalen-2-amine
20571
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P264b-P270-P280-P301+P312-P305+P351+P338-P310-P330-P501c

H302-H318

RTECSNumber : DU3090000

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only