missing translation for 'onlineSavingsMsg'
Learn More

1-(1-Benzothiophen-2-yl)-2-bromo-1-ethanone, ≥97%, Thermo Scientific™

Chemical Identifiers

CAS 97511-06-3
Molecular Formula C10H7BrOS
Molecular Weight (g/mol) 255.13
MDL Number MFCD06658971
InChI Key WVTSOGFICBVCRE-UHFFFAOYSA-N
Synonym 1-1-benzothiophen-2-yl-2-bromo-1-ethanone, 1-1-benzothiophen-2-yl-2-bromoethanone, 2-bromoacetyl benzo b thiophene, 1-1-benzothiophen-2-yl-2-bromoethan-1-one, 1-benzo b thiophen-2-yl-2-bromoethanone, 1-benzo b thiophen-2-yl-2-bromoethan-1-one, 1-benzothiophen-2-yl-2-bromoethan-1-one, 2-bromoacetyl thianaphthene
PubChem CID 2795258
IUPAC Name 1-(1-benzothiophen-2-yl)-2-bromoethanone
SMILES BrCC(=O)C1=CC2=CC=CC=C2S1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
CC12205CB
View Documents Promotion Details
N/A 250mg Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

97511-06-3
255.13
WVTSOGFICBVCRE-UHFFFAOYSA-N
2795258
BrCC(=O)C1=CC2=CC=CC=C2S1
C10H7BrOS
MFCD06658971
1-1-benzothiophen-2-yl-2-bromo-1-ethanone, 1-1-benzothiophen-2-yl-2-bromoethanone, 2-bromoacetyl benzo b thiophene, 1-1-benzothiophen-2-yl-2-bromoethan-1-one, 1-benzo b thiophen-2-yl-2-bromoethanone, 1-benzo b thiophen-2-yl-2-bromoethan-1-one, 1-benzothiophen-2-yl-2-bromoethan-1-one, 2-bromoacetyl thianaphthene
1-(1-benzothiophen-2-yl)-2-bromoethanone

Specifications

97511-06-3
100.0
C10H7BrOS
1-1-benzothiophen-2-yl-2-bromo-1-ethanone, 1-1-benzothiophen-2-yl-2-bromoethanone, 2-bromoacetyl benzo b thiophene, 1-1-benzothiophen-2-yl-2-bromoethan-1-one, 1-benzo b thiophen-2-yl-2-bromoethanone, 1-benzo b thiophen-2-yl-2-bromoethan-1-one, 1-benzothiophen-2-yl-2-bromoethan-1-one, 2-bromoacetyl thianaphthene
BrCC(=O)C1=CC2=CC=CC=C2S1
255.13
255.13
1-(1-Benzothiophen-2-yl)-2-bromo-1-ethanone
96.5
97%
MFCD06658971
WVTSOGFICBVCRE-UHFFFAOYSA-N
1-(1-benzothiophen-2-yl)-2-bromoethanone
2795258
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS
Documents

Documents