missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-(1-Benzofuran-3-yl)-2-bromo-1-ethanone, 95+%, Thermo Scientific™
Specifications
Specifications
| Assay Percent Range | 95% |
| Quantity | 250mg |
| Formula Weight | 239.07 |
| Percent Purity | 95% |
| Chemical Name or Material | 1-(1-Benzofuran-3-yl)-2-bromo-1-ethanone |