Learn More
1,1'-Azobis(N,N-dimethylformamide), 95%
CAS: 10465-78-8 | C6H12N4O2 | 172.188 g/mol
Supplier: Thermo Scientific Chemicals J6486503
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 1,1'-Azobis(N,N-dimethylformamide) | |
| 112°C | |
| C6H12N4O2 | |
| MFCD00008318 | |
| 1910409 | |
| VLSDXINSOMDCBK-BQYQJAHWSA-N | |
| (3E)-3-(dimethylcarbamoylimino)-1,1-dimethylurea | |
| 5353800 | |
| 172.19 | |
| Powder |
| 10465-78-8 | |
| Yellow | |
| (CH3)2NCON=NCON(CH3)2 | |
| 1 g | |
| diamide, n,n,n',n'-tetramethylazodicarboxamide, tmad, 1,1'-azobis n,n-dimethylformamide, tetramethyldiazenedicarboxamide, diazenedicarboxamide, tetramethyl, n,n,n',n'-tetramethylazoformamide, diazenedicarboxylic acid bis n,n-dimethylamide, n,n,n',n'-tetramethylazobisformamide, tetramethylazodicarboxamide | |
| CN(C)C(=O)N=NC(=O)N(C)C | |
| 172.188 | |
| CHEBI:48963 | |
| 95% |
Chemical Identifiers
| 10465-78-8 | |
| 172.188 | |
| VLSDXINSOMDCBK-BQYQJAHWSA-N | |
| 5353800 | |
| (3E)-3-(dimethylcarbamoylimino)-1,1-dimethylurea |
| C6H12N4O2 | |
| MFCD00008318 | |
| diamide, n,n,n',n'-tetramethylazodicarboxamide, tmad, 1,1'-azobis n,n-dimethylformamide, tetramethyldiazenedicarboxamide, diazenedicarboxamide, tetramethyl, n,n,n',n'-tetramethylazoformamide, diazenedicarboxylic acid bis n,n-dimethylamide, n,n,n',n'-tetramethylazobisformamide, tetramethylazodicarboxamide | |
| CHEBI:48963 | |
| CN(C)C(=O)N=NC(=O)N(C)C |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 233-951-7
RTECSNumber : LQ1041000
TSCA : No
Recommended Storage : Store at -20°C
RUO – Research Use Only