missing translation for 'onlineSavingsMsg'
Learn More

1,1'-Azobis(N,N-dimethylformamide), 95%

Catalog No. AAJ6486506
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6486506 5 g
AAJ6486503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6486506 Supplier Thermo Scientific Chemicals Supplier No. J6486506

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 10465-78-8 | C6H12N4O2 | 172.188 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 10465-78-8
Molecular Formula C6H12N4O2
Molecular Weight (g/mol) 172.188
MDL Number MFCD00008318
InChI Key VLSDXINSOMDCBK-BQYQJAHWSA-N
Synonym diamide, n,n,n',n'-tetramethylazodicarboxamide, tmad, 1,1'-azobis n,n-dimethylformamide, tetramethyldiazenedicarboxamide, diazenedicarboxamide, tetramethyl, n,n,n',n'-tetramethylazoformamide, diazenedicarboxylic acid bis n,n-dimethylamide, n,n,n',n'-tetramethylazobisformamide, tetramethylazodicarboxamide
PubChem CID 5353800
ChEBI CHEBI:48963
IUPAC Name (3E)-3-(dimethylcarbamoylimino)-1,1-dimethylurea
SMILES CN(C)C(=O)N=NC(=O)N(C)C

Specifications

Melting Point 112°C
Color Yellow
Linear Formula (CH3)2NCON=NCON(CH3)2
Quantity 5 g
Beilstein 1910409
Formula Weight 172.19
Percent Purity 95%
Physical Form Powder
Chemical Name or Material 1,1'-Azobis(N,N-dimethylformamide)
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 233-951-7
RTECSNumber LQ1041000
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only