missing translation for 'onlineSavingsMsg'
Learn More

1-(1-Adamantyl)ethylamine Hydrochloride 98.0+%, TCI America™

Catalog No. R00705G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R00705G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R00705G Supplier TCI America Supplier No. R00705G

Chemical Identifiers

CAS 1501-84-4
Molecular Formula C12H22ClN
Molecular Weight (g/mol) 215.77
MDL Number MFCD00072023
InChI Key OZBDFBJXRJWNAV-UHFFFAOYNA-N
Synonym rimantadine hydrochloride, flumadine, rimantadine hcl, meradane, 1-1-adamantyl ethylamine hydrochloride, 1-1-aminoethyl adamantane hydrochloride, alpha-methyl-1-adamantanemethylamine hydrochloride, 1-adamantan-1-yl ethanamine hydrochloride, algirem, meradan
PubChem CID 15165
ChEBI CHEBI:8865
IUPAC Name hydrogen 1-(adamantan-1-yl)ethan-1-amine chloride
SMILES [H+].[Cl-].CC(N)C12CC3CC(CC(C3)C1)C2

Specifications

Melting Point 375°C
Color White
Quantity 5 g
Formula Weight 215.77
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material 1-(1-Adamantyl)ethylamine Hydrochloride
RTECSNumber AU4717000
TSCA No