missing translation for 'onlineSavingsMsg'
Learn More

1-(1-Adamantyl)ethylamine hydrochloride, 98%

Catalog No. p-7035658
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5337706 5 g
AAH5337714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5337706 Supplier Thermo Scientific Chemicals Supplier No. H5337706

CAS: 1501-84-4 | C12H22ClN | 215.77 g/mol

1-(1-Adamantyl)ethylamine hydrochloride is indicated for the prophylaxis and treatment of illness caused by various strains of influenza A virus in adults (17 years and older).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-(1-Adamantyl)ethylamine hydrochloride is indicated for the prophylaxis and treatment of illness caused by various strains of influenza A virus in adults (17 years and older).

Solubility
Freely soluble in water.

Notes
Stable under recommended storage conditions. Store away from oxidizing agents.

Chemical Identifiers

CAS 1501-84-4
Molecular Formula C12H22ClN
Molecular Weight (g/mol) 215.77
MDL Number MFCD00072023
InChI Key OZBDFBJXRJWNAV-UHFFFAOYNA-N
Synonym rimantadine hydrochloride, flumadine, rimantadine hcl, meradane, 1-1-adamantyl ethylamine hydrochloride, 1-1-aminoethyl adamantane hydrochloride, alpha-methyl-1-adamantanemethylamine hydrochloride, 1-adamantan-1-yl ethanamine hydrochloride, algirem, meradan
PubChem CID 15165
ChEBI CHEBI:8865
IUPAC Name 1-(1-adamantyl)ethanamine;hydrochloride
SMILES [H+].[Cl-].CC(N)C12CC3CC(CC(C3)C1)C2

Specifications

Melting Point 375°C
Quantity 5 g
Merck Index 148224
Solubility Information Freely soluble in water.
Formula Weight 215.77
Percent Purity 98%
Chemical Name or Material 1-(1-Adamantyl)ethylamine hydrochloride
Hazard Category Acute toxicity (category 4)
Hazard Statement Harmful if swallowed
RTECSNumber AU4717000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only