missing translation for 'onlineSavingsMsg'
Learn More

1,1,4,7,7-Pentamethyldiethylenetriamine, 98%

Catalog No. AAA1543314
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1543314 25 g
AAA1543322 100 g
AAA1543336 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1543314 Supplier Thermo Scientific Chemicals Supplier No. A1543314
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 3030-47-5 | C9H23N3 | 173.304 g/mol

Used as ligand in NMR study of complexation/aggregation with neopentyllithium1 and in the study of ligand effects on the selectivity of CO2 incorporation into α,ω-diynes

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as ligand in NMR study of complexation/aggregation with neopentyllithium1 and in the study of ligand effects on the selectivity of CO2 incorporation into α,ω-diynes

Solubility
Miscible in water.

Notes
Air & Moisture Sensitive. Store under inert gas. Incompatible with strong oxidizing agents, strong acids.

Chemical Identifiers

CAS 3030-47-5
Molecular Formula C9H23N3
Molecular Weight (g/mol) 173.304
MDL Number MFCD00014876
InChI Key UKODFQOELJFMII-UHFFFAOYSA-N
Synonym pentamethyldiethylenetriamine, 1,1,4,7,7-pentamethyldiethylenetriamine, pmdt, n,n,n',n,n-pentamethyldiethylenetriamine, pmdeta, pmdta, pentamethyldiethylenetriaminek, bis 2-dimethylaminoethyl methyl amine, 2,5,8-trimethyl-2,5,8-triazanonane, n,n',n-pentamethyldiethylenetriamine
PubChem CID 18196
ChEBI CHEBI:39475
IUPAC Name N'-[2-(dimethylamino)ethyl]-N,N,N'-trimethylethane-1,2-diamine
SMILES CN(C)CCN(C)CCN(C)C

Specifications

Melting Point <-20°C
Density 0.83
Boiling Point 199°C
Flash Point 72°C (161°F)
Refractive Index 1.442
Quantity 25 g
UN Number UN2922
Beilstein 1741396
Sensitivity Air and moisture sensitive
Solubility Information Miscible in water.
Formula Weight 173.3
Percent Purity 98%
Chemical Name or Material 1,1,4,7,7-Pentamethyldiethylenetriamine
Show More Show Less
Hazard Category H227-H302-H311-H314-H335
Hazard Statement GHS H Statement
H311-H314-H227-H302
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Combustible liquid.
Harmful if swallowed.
Precautionary Statement P210-P235-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.
EINECSNumber 221-201-1
RTECSNumber IE2100000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only