missing translation for 'onlineSavingsMsg'
Learn More

1,1,4,7,10,10-Hexamethyltriethylenetetramine 98.0+%, TCI America™

Catalog No. H13715ML
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H13715ML 5 mL
H137125ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H13715ML Supplier TCI America Supplier No. H13715ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 3083-10-1
Molecular Formula C12H30N4
Molecular Weight (g/mol) 230.40
MDL Number MFCD00025684
InChI Key DWFKOMDBEKIATP-UHFFFAOYSA-N
Synonym 1,1,4,7,10,10-hexamethyltriethylenetetramine, hexamethyltriethylenetetramine, 2,5,8,11-tetramethyl-2,5,8,11-tetraazadodecane, hmteta, 1,2-ethanediamine, n,n'-bis 2-dimethylamino ethyl-n,n'-dimethyl, n1,n1'-ethane-1,2-diyl bis n1,n2,n2-trimethylethane-1,2-diamine, n,n,n',n,n',n'-hexamethyltriethylenetetramine, triethylenetetramine, 1,1,4,7,10,10-hexamethyl, 1,1,4,7,10,10-hexamethyl-triethylenetetramine, n,n'-bis 2-dimethylamino ethyl-n,n'-dimethylethylenediamine
PubChem CID 76509
IUPAC Name N'-[2-[2-(dimethylamino)ethyl-methylamino]ethyl]-N,N,N'-trimethylethane-1,2-diamine
SMILES CN(C)CCN(C)CCN(C)CCN(C)C

Specifications

Color Yellow
Boiling Point 130°C
Quantity 5 mL
UN Number 2735
Formula Weight 230.40
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material 1,1,4,7,10,10-Hexamethyltriethylenetetramine
EINECSNumber (7)-0005
RTECSNumber KH8587135
TSCA Yes