missing translation for 'onlineSavingsMsg'
Learn More

1,1,4,7,10,10-Hexamethyltriethylenetetramine, 97%

Catalog No. aah5608906
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5608906 5 g
AAH5608903 1 g
AAH5608914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH5608906 Supplier Thermo Scientific Chemicals Supplier No. H5608906
Only null left

CAS: 3083-10-1 | C12H30N4 | 230.40 g/mol

1,1,4,7,10,10-Hexamethyltriethylenetetramine may be used as reagent in the synthesis of ideal linear random copolymers containing both vinyl polymer and polyester units in a single polymer chain.1 1,1,4,7,10,10-Hexamethyltriethylenetetramine complexed with CuBr constitutes catalytic complex, used in the copolymerization of poly[ε-caprolactone] with N,N-dimethylamino-2-ethyl methacrylate monomers by atom-transfer radical polymerization (ATRP).2 It may be used as catalyst in the aqueous surface-initiated-ATRP to grow poly(N,N-dimethylacrylamide) (PDMA).3

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,1,4,7,10,10-Hexamethyltriethylenetetramine may be used as reagent in the synthesis of ideal linear random copolymers containing both vinyl polymer and polyester units in a single polymer chain.1 1,1,4,7,10,10-Hexamethyltriethylenetetramine complexed with CuBr constitutes catalytic complex, used in the copolymerization of poly[ε-caprolactone] with N,N-dimethylamino-2-ethyl methacrylate monomers by atom-transfer radical polymerization (ATRP).2 It may be used as catalyst in the aqueous surface-initiated-ATRP to grow poly(N,N-dimethylacrylamide) (PDMA).3

Solubility
Soluble in water.

Notes
Incompatible with oxidizing agents. Store at room temperature.

Chemical Identifiers

CAS 3083-10-1
MDL Number MFCD00025684

Specifications

Density 0.847
Boiling Point 130°C (11 mmHg)
Flash Point 102°C (216°F)
Molecular Formula C7H6N2O2S
Refractive Index 1.456
Quantity 5 g
UN Number UN3725
Synonym 4-cyanobenzene-1-sulfonamide, p-cyanobenzenesulfonamide, 4-cyanobenzenesulphonamide, benzenesulfonamide, p-cyano, 4-cyano-benzenesulfonamide, benzenesulfonamide, 4-cyano, p-cyanobenzenesulphonamide, benzenesulfonamide, 4-cyano-9ci, 4-sulphamoylbenzonitrile, wln: zswr dcn
Solubility Information Soluble in water.
InChI Key UZECCNDOASGYNH-UHFFFAOYSA-N
SMILES C1=CC(=CC=C1C#N)S(=O)(=O)N
IUPAC Name 4-cyanobenzenesulfonamide
Molecular Weight (g/mol) 182.20
PubChem CID 76566
Formula Weight 230.39
Percent Purity 97%
Chemical Name or Material 1,1,4,7,10,10-Hexamethyltriethylenetetramine
Show More Show Less
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: POLYAMINES, LIQUID, CORROSIVE, N.O.S.
EINECSNumber 221-382-7
RTECSNumber KH8587135
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only