missing translation for 'onlineSavingsMsg'
Learn More

1,1,3-Triphenylpropargyl alcohol, 97%

Catalog No. p-7035735
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5346106 5 g
AAH5346114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5346106 Supplier Thermo Scientific Chemicals Supplier No. H5346106

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1522-13-0 | C21H16O | 284.36 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1522-13-0
Molecular Formula C21H16O
Molecular Weight (g/mol) 284.36
MDL Number MFCD00004449
InChI Key VWRQCJRTHKUVNF-UHFFFAOYSA-N
Synonym 1,1,3-triphenylpropargyl alcohol, 1,1,3-triphenyl-2-propyn-1-ol, diphenyl phenylethynyl carbinol, benzenemethanol, .alpha.-phenyl-.alpha.-phenylethynyl, triphenylprop-2-yn-1-ol, 1,1,3-triphenylpropargylalcohol, 2-propyn-1-ol, 1,1,3-triphenyl, pubchem14025, acmc-209d7i, triphenyl-1-propyne-3-ol
PubChem CID 137058
IUPAC Name 1,1,3-triphenylprop-2-yn-1-ol
SMILES OC(C#CC1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 80°C to 82°C
Quantity 5 g
Formula Weight 284.36
Percent Purity 97%
Chemical Name or Material 1,1,3-Triphenylpropargyl alcohol
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H301-H311-H332-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only