missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,1,3,3-Tetramethyldisiloxane 97.0+%, TCI America™
Supplier: TCI America T1437250ML
Specifications
| 71°C | |
| Colorless | |
| Liquid | |
| C4H12OSi2 | |
| 1993 | |
| KWEKXPWNFQBJAY-UHFFFAOYSA-N | |
| dimethylsilylideneoxonio(dimethyl)silanide | |
| 6327482 | |
| ≥97.0% (GC) |
| 1,1,3,3-Tetramethyldisiloxane | |
| 3277-26-7 | |
| 250 mL | |
| MFCD00008256 | |
| 1,1,3,3-tetramethyldisiloxane, tetramethyldisiloxane, dimethylsilyl ether, bis dimethylsilyl ether, bis dimethylsilyl oxide, 1,3-dihydrotetramethyldisiloxane, sym-tetramethyldisiloxane, disiloxane, tetramethyl, dsstox_cid_30574, dsstox_gsid_52010 | |
| C[Si-](C)[O+]=[Si](C)C | |
| 132.309 | |
| 134.33 |
Chemical Identifiers
| 3277-26-7 | |
| 132.309 | |
| KWEKXPWNFQBJAY-UHFFFAOYSA-N | |
| 6327482 | |
| C[Si-](C)[O+]=[Si](C)C |
| C4H12OSi2 | |
| MFCD00008256 | |
| 1,1,3,3-tetramethyldisiloxane, tetramethyldisiloxane, dimethylsilyl ether, bis dimethylsilyl ether, bis dimethylsilyl oxide, 1,3-dihydrotetramethyldisiloxane, sym-tetramethyldisiloxane, disiloxane, tetramethyl, dsstox_cid_30574, dsstox_gsid_52010 | |
| dimethylsilylideneoxonio(dimethyl)silanide |
Safety and Handling
EINECSNumber : (7)-0477
RTECSNumber : JN1000000
TSCA : Yes