missing translation for 'onlineSavingsMsg'
Learn More

1,1,3,3-Tetramethyldisiloxane 97.0+%, TCI America™

Catalog No. T143725ML
Change view
Click to view available options
Quantity:
25 mL
250 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T143725ML 25 mL
T1437250ML 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T143725ML Supplier TCI America Supplier No. T143725ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 3277-26-7
Molecular Formula C4H12OSi2
Molecular Weight (g/mol) 132.309
MDL Number MFCD00008256
InChI Key KWEKXPWNFQBJAY-UHFFFAOYSA-N
Synonym 1,1,3,3-tetramethyldisiloxane, tetramethyldisiloxane, dimethylsilyl ether, bis dimethylsilyl ether, bis dimethylsilyl oxide, 1,3-dihydrotetramethyldisiloxane, sym-tetramethyldisiloxane, disiloxane, tetramethyl, dsstox_cid_30574, dsstox_gsid_52010
PubChem CID 6327482
IUPAC Name dimethylsilylideneoxonio(dimethyl)silanide
SMILES C[Si-](C)[O+]=[Si](C)C

Specifications

Boiling Point 71°C
Color Colorless
Physical Form Liquid
Quantity 25 mL
UN Number 1993
Formula Weight 134.33
Percent Purity ≥97.0% (GC)
Chemical Name or Material 1,1,3,3-Tetramethyldisiloxane
EINECSNumber (7)-0477
RTECSNumber JN1000000
TSCA Yes