missing translation for 'onlineSavingsMsg'
Learn More

1,1,3,3-Tetramethyldisiloxane, 97%

Catalog No. AC216511000
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC216511000 100 mL Glass bottle
AC216510250 25 mL Glass bottle
AC216515000 500 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC216511000 Supplier Thermo Scientific Chemicals Supplier No. 216511000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 3277-26-7 | C4H14OSi2 | 134.33 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent

Chemical Identifiers

CAS 3277-26-7
Molecular Formula C4H14OSi2
Molecular Weight (g/mol) 134.33
InChI Key KWEKXPWNFQBJAY-UHFFFAOYSA-N
Synonym 1,1,3,3-tetramethyldisiloxane, tetramethyldisiloxane, dimethylsilyl ether, bis dimethylsilyl ether, bis dimethylsilyl oxide, 1,3-dihydrotetramethyldisiloxane, sym-tetramethyldisiloxane, disiloxane, tetramethyl, dsstox_cid_30574, dsstox_gsid_52010
PubChem CID 6327482
IUPAC Name dimethylsilylideneoxonio(dimethyl)silanide
SMILES C[Si-](C)[O+]=[Si](C)C

Specifications

Boiling Point 70.0°C to 71.0°C
Melting Point -78.0°C
Color Colorless
Physical Form Liquid
Quantity 100 mL
Flash Point −20°C
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.3690 to 1.3710
Specific Gravity 0.76
Assay Percent Range 97%
Density 0.7600g/mL
Linear Formula [(CH3)2SiH]2O
Solubility Information Solubility in water: decomposes. Other solubilities: decomposes with alcohol
Formula Weight 134.33
Percent Purity 97%
Chemical Name or Material 1,1,3,3-Tetramethyldisiloxane
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Store in a well-ventilated place.
Keep contai
EINECSNumber 221-906-4

RUO – Research Use Only