missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,1,3,3-Tetramethyldisilazane 97.0+%, TCI America™
$76.43 - $210.63
Chemical Identifiers
| CAS | 15933-59-2 |
|---|---|
| Molecular Formula | C4H13NSi2 |
| Molecular Weight (g/mol) | 131.325 |
| MDL Number | MFCD00025626 |
| InChI Key | GJWAPAVRQYYSTK-UHFFFAOYSA-N |
| Synonym | 1,1,3,3-tetramethyldisilazane, bis dimethylsilyl amine, tetramethyldisilazane, tetramethyl disilazane, tmds, silanamine, n-dimethylsilyl-1,1-dimethyl, disilazane, 1,1,3,3-tetramethyl, c4h15nsi2, n-dimethylsilyl-1,1-dimethylsilylamine, 1,3,3-tetramethyldisilazane |
| PubChem CID | 6327317 |
| IUPAC Name | [(dimethyl-$l^{3}-silanyl)amino]-dimethylsilicon |
| SMILES | C[Si](C)N[Si](C)C |
Chemical Identifiers
| 15933-59-2 | |
| 131.325 | |
| GJWAPAVRQYYSTK-UHFFFAOYSA-N | |
| 6327317 | |
| C[Si](C)N[Si](C)C |
| C4H13NSi2 | |
| MFCD00025626 | |
| 1,1,3,3-tetramethyldisilazane, bis dimethylsilyl amine, tetramethyldisilazane, tetramethyl disilazane, tmds, silanamine, n-dimethylsilyl-1,1-dimethyl, disilazane, 1,1,3,3-tetramethyl, c4h15nsi2, n-dimethylsilyl-1,1-dimethylsilylamine, 1,3,3-tetramethyldisilazane | |
| [(dimethyl-$l^{3}-silanyl)amino]-dimethylsilicon |
Specifications
| 100°C | |
| 15933-59-2 | |
| 5 mL | |
| MFCD00025626 | |
| 1,1,3,3-tetramethyldisilazane, bis dimethylsilyl amine, tetramethyldisilazane, tetramethyl disilazane, tmds, silanamine, n-dimethylsilyl-1,1-dimethyl, disilazane, 1,1,3,3-tetramethyl, c4h15nsi2, n-dimethylsilyl-1,1-dimethylsilylamine, 1,3,3-tetramethyldisilazane | |
| C[Si](C)N[Si](C)C | |
| 131.325 | |
| 133.34 | |
| 1,1,3,3-Tetramethyldisilazane |
| Yellow | |
| Liquid | |
| C4H13NSi2 | |
| 2924 | |
| GJWAPAVRQYYSTK-UHFFFAOYSA-N | |
| [(dimethyl-$l^{3}-silanyl)amino]-dimethylsilicon | |
| 6327317 | |
| ≥97.0% (GC) |
Safety and Handling
TSCA : Yes