missing translation for 'onlineSavingsMsg'
Learn More

1,1,3,3-Tetramethyldisilazane 97.0+%, TCI America™

Catalog No. T083325ML
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T083325ML 25 mL
T08335ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T083325ML Supplier TCI America Supplier No. T083325ML
Only null left

Chemical Identifiers

CAS 15933-59-2
Molecular Formula C4H13NSi2
Molecular Weight (g/mol) 131.325
MDL Number MFCD00025626
InChI Key GJWAPAVRQYYSTK-UHFFFAOYSA-N
Synonym 1,1,3,3-tetramethyldisilazane, bis dimethylsilyl amine, tetramethyldisilazane, tetramethyl disilazane, tmds, silanamine, n-dimethylsilyl-1,1-dimethyl, disilazane, 1,1,3,3-tetramethyl, c4h15nsi2, n-dimethylsilyl-1,1-dimethylsilylamine, 1,3,3-tetramethyldisilazane
PubChem CID 6327317
IUPAC Name [(dimethyl-$l^{3}-silanyl)amino]-dimethylsilicon
SMILES C[Si](C)N[Si](C)C

Specifications

Boiling Point 100°C
Color Yellow
Physical Form Liquid
Quantity 25 mL
UN Number 2924
Formula Weight 133.34
Percent Purity ≥97.0% (GC)
Chemical Name or Material 1,1,3,3-Tetramethyldisilazane