missing translation for 'onlineSavingsMsg'
Learn More

1,1,3,3-Tetramethylbutylamine 98.0+%, TCI America™

Catalog No. p-7136104
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 mL
500 mL
Catalog No. Quantity
T014325ML 25 mL
T0143500ML 500 mL
2 options

Catalog No. T014325ML

Supplier: TCI America T014325ML

Only null left

Chemical Identifiers

CAS 107-45-9
Molecular Formula C8H19N
Molecular Weight (g/mol) 129.247
MDL Number MFCD00008053
InChI Key QIJIUJYANDSEKG-UHFFFAOYSA-N
Synonym tert-octylamine, 1,1,3,3-tetramethylbutylamine, 2-pentanamine, 2,4,4-trimethyl, t-octylamine, 2,4,4-trimethyl-2-pentylamine, butylamine, 1,1,3,3-tetramethyl, 1,1,3,3-tetramethylbutanamine, butylamine, bis 1,3-dimethyl, unii-s84lil883b, 2,4,4-trimethyl-2-pentanamine
PubChem CID 61017
IUPAC Name 2,4,4-trimethylpentan-2-amine
SMILES CC(C)(C)CC(C)(C)N

Specifications

Color Colorless
Boiling Point 140°C
Quantity 25 mL
UN Number 2734
Formula Weight 129.25
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 1,1,3,3-Tetramethylbutylamine
EINECSNumber (2)-0133
RTECSNumber EO3850000
TSCA Yes