missing translation for 'onlineSavingsMsg'
Learn More

1,1,3,3-Tetramethylbutylamine 98.0+%, TCI America™

Catalog No. T014325ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T014325ML 25 mL
T0143500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T014325ML Supplier TCI America Supplier No. T014325ML

Chemical Identifiers

CAS 107-45-9
Molecular Formula C8H19N
Molecular Weight (g/mol) 129.247
MDL Number MFCD00008053
InChI Key QIJIUJYANDSEKG-UHFFFAOYSA-N
Synonym tert-octylamine, 1,1,3,3-tetramethylbutylamine, 2-pentanamine, 2,4,4-trimethyl, t-octylamine, 2,4,4-trimethyl-2-pentylamine, butylamine, 1,1,3,3-tetramethyl, 1,1,3,3-tetramethylbutanamine, butylamine, bis 1,3-dimethyl, unii-s84lil883b, 2,4,4-trimethyl-2-pentanamine
PubChem CID 61017
IUPAC Name 2,4,4-trimethylpentan-2-amine
SMILES CC(C)(C)CC(C)(C)N

Specifications

Color Colorless
Boiling Point 140°C
Quantity 25 mL
UN Number 2734
Formula Weight 129.25
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 1,1,3,3-Tetramethylbutylamine
EINECSNumber (2)-0133
RTECSNumber EO3850000
TSCA Yes