missing translation for 'onlineSavingsMsg'
Learn More

1,1,3,3-Tetramethylbutyl isocyanate, 98%

Catalog No. AAL1870603
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1870603 1 g
AAL1870606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1870603 Supplier Thermo Scientific Chemicals Supplier No. L1870603
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1611-57-0 | C9H17NO | 155.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1611-57-0
Molecular Formula C9H17NO
Molecular Weight (g/mol) 155.24
MDL Number MFCD00209615
InChI Key XOHBENIMDRFUIH-UHFFFAOYSA-N
PubChem CID 407498
IUPAC Name 2-isocyanato-2,4,4-trimethylpentane
SMILES CC(C)(C)CC(C)(C)N=C=O

Specifications

Density 0.859
Boiling Point 170°C to 172°C
Flash Point 51°C (123°F)
Odor Pungent
Refractive Index 1.426
Quantity 1 g
UN Number UN3080
Sensitivity Moisture sensitive
Synonym 1,1,3,3-tetramethylbutyl isocyanate, tert-octyl isocyanate, 2-isocyanato-2,4,4-trimethyl-pentane, pentane, 2-isocyanato-2,4,4-trimethyl, acmc-1bnst, pentane,2-isocyanato-2,4,4-trimethyl, unavailable-see asdi_catno 500009647, 1,1,3,3-tetramethylbutyl isocyanate∼2,4,4-trimethyl-2-pentyl isocyanate
Formula Weight 155.24
Percent Purity 98%
Chemical Name or Material 1,1,3,3-Tetramethylbutyl isocyanate
Show More Show Less
Hazard Category H226-H302-H315-H319-H330-H334-H335
Hazard Statement GHS H Statement
H330-H334-H226-H302-H315-H319-H335
Fatal if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Flammable liquid and vapor.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P284-P285-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P363-P370+P378q-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: II
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only