missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,1,3,3-Tetrabutylurea 98.0+%, TCI America™
$169.63 - $428.73
Chemical Identifiers
| CAS | 4559-86-8 |
|---|---|
| Molecular Formula | C17H36N2O |
| Molecular Weight (g/mol) | 284.488 |
| MDL Number | MFCD00042901 |
| InChI Key | SNDGLCYYBKJSOT-UHFFFAOYSA-N |
| PubChem CID | 20691 |
| IUPAC Name | 1,1,3,3-tetrabutylurea |
| SMILES | CCCCN(CCCC)C(=O)N(CCCC)CCCC |
Chemical Identifiers
| 4559-86-8 | |
| 284.488 | |
| SNDGLCYYBKJSOT-UHFFFAOYSA-N | |
| 1,1,3,3-tetrabutylurea |
| C17H36N2O | |
| MFCD00042901 | |
| 20691 | |
| CCCCN(CCCC)C(=O)N(CCCC)CCCC |
Specifications
| 4559-86-8 | |
| Yellow | |
| C17H36N2O | |
| 5 g | |
| CCCCN(CCCC)C(=O)N(CCCC)CCCC | |
| 284.488 | |
| 284.49 | |
| Liquid |
| -60°C | |
| 163°C | |
| MFCD00042901 | |
| SNDGLCYYBKJSOT-UHFFFAOYSA-N | |
| 1,1,3,3-tetrabutylurea | |
| 20691 | |
| ≥98.0% (GC) | |
| 1,1,3,3-Tetrabutylurea |
Safety and Handling
RTECSNumber : YU2080000
TSCA : Yes