missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,1,3,3-Tetrabromoacetone 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 38°C |
| Color | White-Yellow |
| Quantity | 25 g |
| Formula Weight | 373.66 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 1,1,3,3-Tetrabromoacetone |
Safety and Handling
| TSCA | No |
| Recommended Storage | Freezer |