missing translation for 'onlineSavingsMsg'
Learn More

1,1,3,3-Tetrabromoacetone 98.0+%, TCI America™

Catalog No. p-7136559
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T10265G 5 g
T102625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T10265G Supplier TCI America Supplier No. T10265G

Chemical Identifiers

CAS 22612-89-1
Molecular Formula C3H2Br4O
Molecular Weight (g/mol) 373.664
MDL Number MFCD00059470
InChI Key SAMNBOHOBWEEEU-UHFFFAOYSA-N
Synonym 1,1,3,3-Tetrabromo-2-propanone
PubChem CID 608441
IUPAC Name 1,1,3,3-tetrabromopropan-2-one
SMILES C(C(=O)C(Br)Br)(Br)Br

Specifications

Melting Point 38°C
Color White-Yellow
Quantity 5 g
Formula Weight 373.66
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,1,3,3-Tetrabromoacetone
TSCA No
Recommended Storage Freezer