missing translation for 'onlineSavingsMsg'
Learn More

1,1,3,3,5,5,7,7,9,9,11,11,13,13-Tetradecamethylheptasiloxane 95.0+%, TCI America™

Catalog No. T35485ML
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T35485ML 5 mL
T35481ML 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T35485ML Supplier TCI America Supplier No. T35485ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 19095-23-9
Molecular Formula C14H44O6Si7

Specifications

Color Colorless
Boiling Point 128°C
Quantity 5 mL
Formula Weight 505.10
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,1,3,3,5,5,7,7,9,9,11,11,13,13-Tetradecamethylheptasiloxane