missing translation for 'onlineSavingsMsg'
Learn More

1,1,2-Trimethyl-1H-benz[e]indole, 96%

Catalog No. AC421900250
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC421900250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC421900250 Supplier Thermo Scientific Chemicals Supplier No. 421900250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 41532-84-7 | C15H15N | 209.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

CAS 41532-84-7
CAS Min % 95.0
CAS Max % 100.0
Melting Point 111°C to 117°C
Color Yellow to Brown
Density 0.7000g/mL
Infrared Spectrum Authentic
Assay Percent Range 95% min. (HPLC)
Packaging Glass bottle
Molecular Formula C15H15N
Quantity 25 g
MDL Number MFCD00082627
Specific Gravity 0.7
Synonym 1,1,2-trimethyl-1h-benzo e indole, 1,1,2-trimethyl-1h-benz e indole, 1,1,2-trimethylbenz e indole, 2,3,3-trimethyl-4,5-benzo-3h-indole, 2,3,3-trimethylbenzoindolenine, 1,1,2-trimethylbenzo e indole, 1h-benz e indole, 1,1,2-trimethyl, 2,3,3-trimethyl-1h-benz e indolenine, 1,1,2-trimethylhydrobenzo e indole
Solubility Information Solubility in water: insoluble (20°C). Other solubilities: soluble in methanol,acetonitrile (20°C)
InChI Key WJZSZXCWMATYFX-UHFFFAOYSA-N
SMILES CC1=NC2=CC=C3C=CC=CC3=C2C1(C)C
IUPAC Name 1,1,2-trimethyl-1H-benzo[e]indole
Molecular Weight (g/mol) 209.29
PubChem CID 170530
Formula Weight 209.29
Percent Purity 96%
Physical Form Crystalline Powder
Chemical Name or Material 1, 1, 2-Trimethyl-1H-benz[e]indole, 96%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 255-429-8
TSCA TSCA

RUO – Research Use Only