missing translation for 'onlineSavingsMsg'
Learn More

1,1'-(2,6-Pyridinediyl)bis(3-methylimidazolium) Dibromide 98.0+%, TCI America™

Catalog No. p-7135268
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P18161G 1 g
P18165G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P18161G Supplier TCI America Supplier No. P18161G
Only null left

Chemical Identifiers

CAS 263874-05-1
Molecular Formula C13H15Br2N5
Molecular Weight (g/mol) 401.11
MDL Number MFCD11975436
InChI Key VAESRVWVZJLVIA-UHFFFAOYSA-L
PubChem CID 11235002
IUPAC Name 3-methyl-1-[6-(3-methyl-1H-imidazol-3-ium-1-yl)pyridin-2-yl]-1H-imidazol-3-ium dibromide
SMILES [Br-].[Br-].C[N+]1=CN(C=C1)C1=CC=CC(=N1)N1C=C[N+](C)=C1

Specifications

Quantity 1 g
Formula Weight 401.11
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1,1′-(2,6-Pyridinediyl)bis(3-methylimidazolium) Dibromide