missing translation for 'onlineSavingsMsg'
Learn More

1,1,2,2-Tetraphenyldisilane 97.0+%, TCI America™

Catalog No. T18965G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T18965G 5 g
T18961G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T18965G Supplier TCI America Supplier No. T18965G
Only null left

Chemical Identifiers

CAS 16343-18-3
Molecular Formula C24H20Si2
Molecular Weight (g/mol) 364.594
MDL Number MFCD00093170
InChI Key BXLNWOAYQXBHCY-UHFFFAOYSA-N
Synonym TPDS
PubChem CID 10948558
IUPAC Name diphenylsilylidene(diphenyl)silane
SMILES C1=CC=C(C=C1)[Si](=[Si](C2=CC=CC=C2)C3=CC=CC=C3)C4=CC=CC=C4

Specifications

Melting Point 80°C
Quantity 5 g
Formula Weight 366.61
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,1,2,2-Tetraphenyldisilane