Learn More
1,1,2,2-Tetramethyl-1,2-diphenyldisilane, 95%
CAS: 1145-98-8 | C16H22Si2 | 270.522 g/mol
$125.16 - $344.23
Chemical Identifiers
| CAS | 1145-98-8 |
|---|---|
| Molecular Formula | C16H22Si2 |
| Molecular Weight (g/mol) | 270.522 |
| MDL Number | MFCD00054786 |
| InChI Key | IIOOIYHUTINYQO-UHFFFAOYSA-N |
| Synonym | 1,2-diphenyltetramethyldisilane, 1,1,2,2-tetramethyl-1,2-diphenyldisilane, disilane,1,1,2,2-tetramethyl-1,2-diphenyl, 1,2-diphenyl-1,1,2,2-tetramethyldisilane, tetramethyldiphenyldisilan, amtsi083, tetramethyldisilane-1,2-diyldibenzene, bisphenyl-1,1,2,2-tetramethyldisilane, dimethyl phenyl silyl-dimethyl-phenylsilane, 1,1,2,2-tetramethyldiphenyldisilane |
| PubChem CID | 136916 |
| IUPAC Name | [dimethyl(phenyl)silyl]-dimethyl-phenylsilane |
| SMILES | C[Si](C)(C1=CC=CC=C1)[Si](C)(C)C2=CC=CC=C2 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH2870503
|
Thermo Scientific Chemicals
H2870503 |
1 g |
Each for $125.16
|
|
|||||
|
AAH2870506
|
Thermo Scientific Chemicals
H2870506 |
5 g |
Each for $344.23
|
|
|||||
Description
Dimethylphenylsilane was catalytically dehydrogenated and condensed in the presence of platinum complexes to give 1,1,2,2-tetramethyl-1,2-diphenyl-disilane.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsDimethylphenylsilane was catalytically dehydrogenated and condensed in the presence of platinum complexes to give 1,1,2,2-tetramethyl-1,2-diphenyl-disilane.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Chemical Identifiers
| 1145-98-8 | |
| 270.522 | |
| IIOOIYHUTINYQO-UHFFFAOYSA-N | |
| 136916 | |
| C[Si](C)(C1=CC=CC=C1)[Si](C)(C)C2=CC=CC=C2 |
| C16H22Si2 | |
| MFCD00054786 | |
| 1,2-diphenyltetramethyldisilane, 1,1,2,2-tetramethyl-1,2-diphenyldisilane, disilane,1,1,2,2-tetramethyl-1,2-diphenyl, 1,2-diphenyl-1,1,2,2-tetramethyldisilane, tetramethyldiphenyldisilan, amtsi083, tetramethyldisilane-1,2-diyldibenzene, bisphenyl-1,1,2,2-tetramethyldisilane, dimethyl phenyl silyl-dimethyl-phenylsilane, 1,1,2,2-tetramethyldiphenyldisilane | |
| [dimethyl(phenyl)silyl]-dimethyl-phenylsilane |
Specifications
| 111°CC | |
| 1145-98-8 | |
| C16H22Si2 | |
| 1,2-diphenyltetramethyldisilane, 1,1,2,2-tetramethyl-1,2-diphenyldisilane, disilane,1,1,2,2-tetramethyl-1,2-diphenyl, 1,2-diphenyl-1,1,2,2-tetramethyldisilane, tetramethyldiphenyldisilan, amtsi083, tetramethyldisilane-1,2-diyldibenzene, bisphenyl-1,1,2,2-tetramethyldisilane, dimethyl phenyl silyl-dimethyl-phenylsilane, 1,1,2,2-tetramethyldiphenyldisilane | |
| IIOOIYHUTINYQO-UHFFFAOYSA-N | |
| [dimethyl(phenyl)silyl]-dimethyl-phenylsilane | |
| 136916 | |
| 97% |
| 34°C to 37°C | |
| 1 g | |
| MFCD00054786 | |
| Insoluble in water. | |
| C[Si](C)(C1=CC=CC=C1)[Si](C)(C)C2=CC=CC=C2 | |
| 270.522 | |
| 270.52 | |
| 1,1,2,2-Tetramethyl-1,2-diphenyldisilane |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only