missing translation for 'onlineSavingsMsg'
Learn More

1,1,2,2-Tetramethyl-1,2-diphenyldisilane, 95%

Catalog No. AAH2870506
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2870506 5 g
AAH2870503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2870506 Supplier Thermo Scientific Chemicals Supplier No. H2870506
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1145-98-8 | C16H22Si2 | 270.522 g/mol

Dimethylphenylsilane was catalytically dehydrogenated and condensed in the presence of platinum complexes to give 1,1,2,2-tetramethyl-1,2-diphenyl-disilane.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dimethylphenylsilane was catalytically dehydrogenated and condensed in the presence of platinum complexes to give 1,1,2,2-tetramethyl-1,2-diphenyl-disilane.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Chemical Identifiers

CAS 1145-98-8
Molecular Formula C16H22Si2
Molecular Weight (g/mol) 270.522
MDL Number MFCD00054786
InChI Key IIOOIYHUTINYQO-UHFFFAOYSA-N
Synonym 1,2-diphenyltetramethyldisilane, 1,1,2,2-tetramethyl-1,2-diphenyldisilane, disilane,1,1,2,2-tetramethyl-1,2-diphenyl, 1,2-diphenyl-1,1,2,2-tetramethyldisilane, tetramethyldiphenyldisilan, amtsi083, tetramethyldisilane-1,2-diyldibenzene, bisphenyl-1,1,2,2-tetramethyldisilane, dimethyl phenyl silyl-dimethyl-phenylsilane, 1,1,2,2-tetramethyldiphenyldisilane
PubChem CID 136916
IUPAC Name [dimethyl(phenyl)silyl]-dimethyl-phenylsilane
SMILES C[Si](C)(C1=CC=CC=C1)[Si](C)(C)C2=CC=CC=C2

Specifications

Boiling Point 111°CC
Melting Point 34°C to 37°C
Quantity 5 g
Solubility Information Insoluble in water.
Formula Weight 270.52
Percent Purity 97%
Chemical Name or Material 1,1,2,2-Tetramethyl-1,2-diphenyldisilane
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only