missing translation for 'onlineSavingsMsg'
Learn More

1,1,2,2-Tetrafluoroethyl 2,2,2-Trifluoroethyl Ether 99.0+%, TCI America™

Catalog No. T30575G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T30575G 5 g
T305725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T30575G Supplier TCI America Supplier No. T30575G
Only null left

Chemical Identifiers

CAS 406-78-0
Molecular Formula C4H3F7O
Molecular Weight (g/mol) 200.056
MDL Number MFCD00236118
InChI Key CWIFAKBLLXGZIC-UHFFFAOYSA-N
PubChem CID 164596
IUPAC Name 1,1,2,2-tetrafluoro-1-(2,2,2-trifluoroethoxy)ethane
SMILES C(C(F)(F)F)OC(C(F)F)(F)F

Specifications

Color Yellow
Boiling Point 56°C
Quantity 5 g
UN Number 3271
Formula Weight 200.06
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,1,2,2-Tetrafluoroethyl 2,2,2-Trifluoroethyl Ether
EINECSNumber (2)-3983
TSCA Yes